About this item
The HP-3G10 antibody is specific for human CD161, also known as NKR-P1A, a type II transmembrane lectin-like receptor and member of the killer cell lectin-like receptor (KLR) family. CD161 exists as a homodimer which is prominently expressed on natural killer (NK) and NKT cells, where it is proposed to regulate the function of both cell types. CD161 is also found on T cell subsets, including T regulatory cells (Tregs), memory/effector CD4+ T cells, and CD8+ T cells. Th17 cells have been demonstrated to co-express CD161, as surface IL-17A+ cells are contained within the CD161+ fraction of CD4 T cells, so that CD161 (in combination with CCR6) is often used as a marker for enrichment of Th17 cells.
The HP-3G10 antibody may be used for flow cytometric analysis of CD161 on NK and NKT cells, as well as on various T cell subsets. The antibody is also reported to be cross-reactive with Baboon, Chimpanzee and Rhesus CD161.
Type: Primary
Antigen: KLRB1
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee, Baboon, Rhesus
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10051-842
- 10051-840
- Antigen name:
- Killer cell lectin-like receptor subfamily B, member 1
- Killer cell lectin-like receptor subfamily B, member 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HP-3G10
- HP-3G10
- Host:
- Mouse
- Mouse
- Gene ID:
- 3820
- 3820
- Isotype:
- IgG1
- IgG1
Specifications
Related Information
tds-75-1619.pdf
Datasheet
Tonbo Biosciences violetFluor™ Antibodies.pdf