About this item
The GARP5 antibody reacts with human GARP, also known as LRRC32 or Garpin, an 80 kDa transmembrane protein which acts as a receptor for the latent form of TGF-beta 1 (pro-TGF-beta), preventing its secretion. Specifically, GARP is reported to associate with the pro-domain of TGF-beta 1, known as latency-associated peptide (LAP), which must be cleaved to release the biologically active cytokine. The putative role of GARP in sequestering the TGF-beta is important for regulating its activity, as TGF-beta signaling is involved in development and function of Th17, Treg and many other immune cell types. GARP expression has been shown on platelets and at high levels on Foxp3+ Treg cells, where it is proposed to be a phenotypic identifier for activated human Treg cells.
The GARP5 antibody may be used for analysis of human GARP by flow cytometry, immunoprecipitation and Western blotting. Please choose the appropriate format for each application.
Type: Primary
Antigen: LRRC32
Clonality: Monoclonal
Clone: GARP5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10051-394
- 10051-396
- Antigen name:
- Leucine Rich Repeat Containing 32
- Leucine Rich Repeat Containing 32
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- GARP5
- GARP5
- Host:
- Mouse
- Mouse
- Gene ID:
- 2615
- 2615
- Isotype:
- IgG1
- IgG1