Specifications
- Description:Tissue cassette storage cabinet
- Material:Epoxy powder-coated steel
- Cat. no.:10144-720
- Supplier No.:143000
Specifications
About this item
A robust, modular storage solution for formalin fixed paraffin embedded tissue samples.
- Economical option for storing cassettes
- Foam organizers included
- Easy slide drawers with customizable labels
Each drawer includes easily removable crystal clear trays along with foam organizers for keeping cassettes upright.
Store up to 1200 cassettes in the easy slide drawers, each with customizable labels. The non-tip design allows for stacking of up to 3 cabinets high.