About this item
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.
The HI30 antibody is widely used as a marker for human CD45 expression on T cells, B cells, monocytes, macrophages, and NK cells.
Type: Primary
Antigen: PTPRC
Clonality:
Clone: HI30
Conjugation: redFluor® 710
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10141-116
- 10141-114
- Antigen name:
- Protein Tyrosine Phosphatase Receptor Type C
- Protein Tyrosine Phosphatase Receptor Type C
- Antibody type:
- Primary
- Primary
- Clone:
- HI30
- HI30
- Host:
- Mouse
- Mouse
- Gene ID:
- 5788
- 5788
- Isotype:
- IgG1
- IgG1
- Neutralisation:
- Yes
- Yes