About this item
The RA3-6B2 antibody reacts with the human and mouse CD45 isoform known as CD45R, or B220, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns, all critical for leukocyte function. In mouse, the CD45R/B220 isoform is predominantly found on B cells, at varying levels on all stages from pro-B cells to activated B cells, and may also be detected on certain T cell and NK cell subsets. It is of note that B220 is not similarly expressed on human B cells, where it appears to be differentiation-specific and therefore expressed on only some B cell subsets. Other forms of CD45 with restricted cellular expression include CD45RA, CD45RB and CD45RO.
The RA3-6B2 antibody is one of the most consistently used leukocyte markers for B cells, T cell subsets and NK cell subsets in human and mouse. This antibody has also been reported as cross-reactive with feline CD45R/B220.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Cat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76448-896
- 10050-760
- 10050-762
- 10050-758
- Antigen name:
- Protein Tyrosine Phosphatase Receptor Type C
- Protein Tyrosine Phosphatase Receptor Type C
- Protein Tyrosine Phosphatase Receptor Type C
- Protein Tyrosine Phosphatase Receptor Type C
- Antibody type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- RA3-6B2
- RA3-6B2
- RA3-6B2
- RA3-6B2
- Host:
- Rat
- Rat
- Rat
- Rat
- Gene ID:
- 19264
- 19264
- 19264
- 19264
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a