VISION 10™ Polyester Knit Cleanroom Wipers, High-Tech Conversions
Supplier: High-Tech Conversions
Certificates
About this item
VISION 10™ wipers are extremely low in particle generation and chemical extractables. The fabric consists of premium quality virgin polyester fibers that are continuously filamented throughout the knit construction.
- Lint-free
- Cleanroom class 100 (ISO Class 5)
- Cleanroom laundered
- Polyester
- Individually inspected
- Abrasion resistant
- Absorbant
VISION 10™ wipers are knitted on state-of-the-art equipment especially for use in cleanrooms. This fabric undergoes a multitude of special processes that include cleaning, scouring, cutting and a special proprietary cleaning process.
Most importantly, VISION 10™ wipers are laundered in a class 100 cleanroom using ultra-filtered water of 0.45 µ purity and HEPA-filtered air during moisture removal.
All VISION 10™ wipers are individually inspected with a lot-to-lot traceability and using ISO 9000 standards.
Ordering information: Wipers are packaged either 150 or 500 per bag, double-bagged, with 10 bags per case.
Specifications
- Catalog No:
- 10141-364
- 10141-362
- Basis weight:
- 115 g/m²
- 115 g/m²