Specifications
- Description:PermaBlue/HRP
- Size:30 ml
- Cat. no.:CA10064-840
Specifications
About this item
PermaBlue/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase
PermaBlue/HRP can be permanently mounted to produce a strong azure blue color that can be easily distinguished from other stains.