Pipettes, Micro Capillary, TLC Spotting, Ace Glass Incorporated
Catalog #: 10099-066
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Length:5"
- Description:Micro Capillary Pipets
- Cat. no.:10099-066
- Supplier No.:5922-10
Specifications
About this item
Uncalibrated, useful for spotting both aqueous and organic solutions.
Sturdy construction helps hold breakage to a minimum. 5" length. Supplied 100 tubes per clear plastic vial.
Customers Who Bought This Also Bought