Sodium nitrate
Catalog #: 80705-023
Supplier: Avantor
CAS Number: 7631-99-4
Specifications
- Packaging:Bag
- Size:22.7 kg
- Cat. no.:80705-023
- Supplier No.:AMASMR117-22.7KG
Specifications
About this item
Boiling Pt:380 °C (1013 hPa)
Melting Pt:308 °C
Density:2.26 g/cm³ (20 °C)
ADR:5.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 380 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 308 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.99 g/mol
Merck Index:12,08792
Formula:NaNO₃
MW:84.99 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00011119
CAS Number:7631-99-4
EINECS:231-554-3
UN:1498
REACH:01-2119488221-41
:5.1
:III
:GHS03
:GHS07
Customers Who Bought This Also Bought