Sodium carbonate (Soda ash 58%, Light) light
Catalog #: 80705-009
Supplier: Avantor
CAS Number: 497-19-8
Specifications
- Packaging:Bag
- Size:25 kg
- Cat. no.:80705-009
- Supplier No.:AMAS029-945-01
Specifications
About this item
Synonyms: Calcined soda
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 854 °C
:Flash Pt:
:Flash Pt:
:M.W. 105.99 g/mol
Merck Index:14,08596
Formula:Na₂CO₃
MW:105.99 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003494
CAS Number:497-19-8
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
Customers Who Bought This Also Bought