Borax, anhydrous
Catalog #: 80705-012
Supplier: Avantor
CAS Number: 1330-43-4
Specifications
- Packaging:Bag
- Size:25 kg
- Cat. no.:80705-012
- Supplier No.:AMAS029-940-02
Specifications
Specification Test Results
- B₂O₃:68.5 to 69.4%
- Na₂B₄O₇:99.98 to 100.31%
- Na₂O:30.5 to 30.91%
- H₂O:0 - 1.02%
- Fe:0 - 0040%
- Borax, anhydrous:
Specification Test Results
About this item
Synonyms: Borax
:Length:
:Width:
:Height:
:Ø
:Thickness:
:12 Mesh
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 742 °C
:Flash Pt:
:Flash Pt:
:M.W. 201.22 g/mol
:Borax
:742
:1575
:GHS08
:12,08733
Formula:Na₂B₄O₇
MW:201.22 g/mol
MDL Number:MFCD00081185
CAS Number:1330-43-4
Customers Who Bought This Also Bought