About this item
ECO current replacement for Ecoline
The intuitive menu navigation allows for easy operation. Both control heads are equipped with a mini-USB interface as a standard feature. Additional interfaces are available via the add-on modules. On-the-fly adjustment of the flow between internal and external circulation is made possible even during operation via the new flow rate switch. The more powerful units are equipped with the energy-saving LAUDA SmartCool system. All cooling thermostats are available with air-cooled or water-cooled options. Stainless steel bath.
There are innovations and developments particularly with regard to the range of features and the ease of operation. Both control heads, designated ECO Silver and ECO Gold, have a powerful circulating pump with more than 30 percent higher pump capacity compared to the predecessor models.
Specifications
- Catalog No:
- 89202-394
- 89202-388
- 89202-396
- 89202-478
- 89202-400
- 89202-488
- 89202-398
- 89202-486
- 89202-484
- 89202-408
- 89202-406
- 89202-492
- 89202-402
- 89202-490
- 89202-412
- 89202-410
- 89202-502
- 89202-500
- 89202-498
- 89202-504
- 89202-386
- Pump flow rate:
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- 22 L/min. (Max.)
- Pump pressure:
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)
- 0.55 Bar (Max.)