Order Entry
ContactUsLinkComponent
VWR® LB Broth Liquid
VWR® LB Broth Liquid
Catalog #: 89500-596
Supplier:  VWR
CAS Number:  
Lb Broth Liquid
VWR® LB Broth Liquid
Catalog #: 89500-596
Supplier:  VWR
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 L
  • Storage conditions:
    Room temperature
  • Cat. no.:
    89500-596
  • Supplier No.:
    J833-1L

Specifications

About this item

LB Broth, Liquid is used for the propagation and maintenance of E.coli. Widely used for the preparation of plasmid DNA and recombinant proteins.

  • Ready to use liquid media
  • Clear, yellow to brown liquid

1L of media contains 10 grams of sodium chloride, 10 grams of tryptone/ cesein peptone, 5 grams of yeast extract bacteriological.

No growth detected in Bioburden (Milliflex) (EQC) test. pH @25°C is 6.65 - 7.15. It has refractive index of 1.334 - 1.342. Specific Gravity at 20°C is 1.0116 - 1.0128. Abs.@260nm (1:100, Water) is 0.2263 - 0.2663.

Customers Who Bought This Also Bought