Safety Funnel with Ball Valve, S.C.A.T.
Supplier: S.C.A.T.
Certificates
About this item
Made of high quality PE-HD for use with all types of chemicals.
- Splash protection allows uniform discharge without splashes
- The ball valve floats and automatically closes after filling
For the disposal of small quantities and sticky liquid waste into the container, we recommend to rinse enough with water after using the funnel with ball valve, to prevent a potential sticking or stucking of the ball valve. Freely rotatable cap comes in different thread sizes for a wide variety of containers. Safety lance ensures clean outflow without contaminating the container opening. With electrically conductive funnels (black), the lance facilitates a safe additional grounding of the contents. Models in black are made of electrically conductive PE-HD and have a ground connection to prevent sparking hazards.
BodyTextAccessories: Sieves are for trapping stirrers or larger contaminant particles.
Customers Who Bought This Also Bought