ContactUsLinkComponent
VWR® Vacuum Oven Gasket
VWR® Vacuum Oven Gasket
Catalog #: 14231-996
Supplier:  VWR International
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
VWR® Vacuum Oven Gasket
Catalog #: 14231-996
Supplier:  VWR International
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Dimensions:
    9 x 9"
  • Description:
    Gasket
  • Material:
    Fluorosilicone
Specifications
About this item
For use with VWR® Vacuum Ovens.
  • Temperature-resistant construction
  • Resilient against wide changes in pressure
  • Fluorosilicone fabrication
Gaskets are designed for specific applications with vacuum ovens.
Max. temperature: 175 °C
Customers Who Bought This Also Bought