Specifications
- Weight:68 kg
- Description:Mines Assay Sample Drying Oven, 5 Shelf, 240V, 5000W, 3 PH, 60HZ, 3 x 1500W Elements
- L×W×H:97 x 48 x 183 cm
- Cat. no.:CA11019-598
- Supplier No.:SP107
Specifications
About this item
This oven is ideal for demanding laboratory analysis. Designed for temperatures up to 150°C, its sturdy design features a 5cm bottom reinforcing Z bar, 10cm heavy-duty castors, key-lock door handles, and its body and doors are covered with 5cm fiberglass insulation to save energy and money. It also features an advanced, dependable electrical design with a caloritech thermostat, inside temperature probe, 110V (50 C.F.M.) fan with separate on/off switch, main fused disconnect, and S.E.W.-II wiring. The oven is Ontario Hydro approved, ensuring dependability and safe operation. Available in two voltages. Convenient and versatile, it comes with five adjustable shelves (90x42cm) but can accommodate 10 total. Additional shelves purchased separately.
- Energy efficient
- Heavy-duty design
- Adjustable shelving
- Range of sizes/voltages