Hot Crucible Tray
Catalog #: CA11019-504
Supplier: G-L LEBEL CA
Specifications
- Cat. no.:CA11019-504
- L×W×D:56 x 38 x 10 cm (22 x 15 x 4")
- Supplier No.:SP109F
Specifications
About this item
The hot crucible tray is specially designed for carrying hot fire assay crucibles.
Its 3.2mm mild steel sides and heavy wire mesh bottom ensure durability and long life. For convenience and ease of use, it is hinged on one end, allowing placement of crucibles on the tray with crucible tongs after a fused sample has been poured into mold.
Ordering information: Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought