Wooden Button Tray
Catalog #: CA11019-500
Supplier: G-L LEBEL CA
Specifications
- Description:Wooden Button Tray
- Cat. no.:CA11019-500
- Supplier No.:FA125
Specifications
About this item
The button tray is specially designed for carrying lead buttons from fire assay fusions
It is made from 19mm plywood, with holes recessed 6.3mm, and holds 30 buttons.
Ordering information: Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought