To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 mg
- Storage conditions:Store at 4 °C
- Protein/peptide name:PERCP
- Tested applications:SMCC Activated Peridini Chlorophyll Complex (PerCP)
- Cat. no.:10065-510
- Supplier No.:D11-000
Specifications
About this item
Peridinin is a light-harvesting pigment associated with chlorophyll and found in the peridinin chlorophyll binding protein (PerCP); a light harvesting protein found in dinoflagellates. The PerCP complex has a high ratio of peridinin to chlorophyll; most light-harvesting complexes contain more chlorophyll than carotenoids, but the PerCP contains eight peridinin and two chlorophyll molecules arranged to promote peridinin-chlorophyll energy transfer. The PerCP complex is a trimer with a molecular weight of 35kDa.
- Emission: 676 nm
- Excitation: 483 nm