About this item
Hach Pocket Pro is engineered to deliver accurate results.
- Confidence in results
- Save time and money
- Ease of use
With calibration diagnostics. Manual langauges include English, French, German, Italian, and Spanish. No backlight. No replaceable sensor.
Backed up with built in performance diagnostics, you never have to guess when to clean or calibrate the sensor. You also get replaceable batteries for convenient field use, and a large, easy-to-read LCD screen.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10065-860
- pH range:
- 0.0 - 14.0
- Battery life:
- 200
- Power supply:
- Batteries
- Operating temperature range:
- 0…50 °C
- IP rating:
- IP 67
- Parameters:
- pH, Temp
- Calibration:
- 3 point