Graduated Reusable Pipettes, Dynalon
Supplier: Dynalon
Certificates
About this item
Graduated polypropylene pipettes are translucent and resistant to breakage.
- Blue graduations
- Ring mark
- Inscriptions are standard on all pipettes
Ideal for fluoro and silicate chemistry. Recommended cleaning with mild alkaline detergent up to 60 °C (140 °F).
Note: Exposure to temperatures above 60°C may cause volume changes.
Customers Who Bought This Also Bought