Batch Pressure Filters, Sepor
Supplier: SEPOR CA
Certificates
About this item
The lid seals with a rubber gasket, while the cylinder bottom seals with a rubber gasket, while the cylinder bottom seals against pre-wetted filter paper and cloth on the base. Sepor's exclusive HI-SEAL filter cloth should be used when maximum liquid recovery is required. This cloth has a rubber perimeter, and eliminates leakage.
This economical batch pressure filter features a self-supporting lid and yoke assembly that tilts back 30°, allowing the cylinder to be removed between batches. The steel base has a machined waffle grid drainage pattern and bottom drain for bench mounting, or a side drain for floor mounting.
Ordering information: Included are a pressure gauge, safety relief valve, bleed valve, quick-connect air coupling, filter cloth, HI-SEAL filter cloth, package of 100 wet strength filter papers, and a rubber gasket.
Customers Who Bought This Also Bought