Crucible Wood Tray, Anachemia
Catalog #: CA11019-502
Supplier: G-L LEBEL CA
Specifications
- Description:Trays for 30/40 g Crucibles
- Cat. no.:CA11019-502
- Supplier No.:029-810-06
Specifications
About this item
The crucible wood tray is specially designed for carrying crucibles for fire assay fusions.
- Tray consists of a sturdy 16 mm plywood base with oak sides
- Accommodates 24 crucibles (in a 4 x 6 format)
Tray dimensions: 56L x 40W x 10H cm
Customers Who Bought This Also Bought