B.O.D Disposable Bottles, Environmental Express
Supplier: Avantor
Certificates
About this item
Lightweight and disposable bottles manufactured from oriented PET plastic materials.
- Each 300 ml B.O.D
Stoppers are made from unbreakable acrylic.
Bottle is coated with amorphous carbon to seal the bottle. This carbon coating prevents the diffusion of oxygen into or out of the bottle. Clean bottles eliminate the problem o fcontamination from sample carry-over or detergent residue. Bottles have been laboratory tested and found to provide equivalent results to glass bottles.
Customers Who Bought This Also Bought