Vikan Wall Wash Brushes, Remco Products
Supplier: Remco Products
Certificates
About this item
This deck scrub has very effective water distribution and is ideal for use in heavily soiled areas such as meat processing plants.
- Dishwasher safe
- Autoclavable up to 250°F
Block material is polypropylene; bristle material is polyester.
Width: 3"
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought