Porcelain Crucible Marking Ink, CoorsTek
Catalog #: 36085-005
Supplier: Coors Ceramics
Specifications
- Description:Marking Ink, 15 mL
- Cat. no.:36085-005
- Supplier No.:60015
Specifications
About this item
Porcelain Crucible Marking Ink is designed for permanently marking laboratory and other ceramic ware.
- May be applied with pen or brush
- Marked surface must be heat-treated to become permanent
- Instructions are provided on the bottle
Customers Who Bought This Also Bought