Order Entry
ContactUsLinkComponent
Recombinant Endog Fragment (from E. coli)
Recombinant Endog Fragment (from E. coli)
Catalog #: 10073-124
Supplier:  Prosci
CAS Number:  
EndoG Fragment Recombinant Protein Host: E.coli, EndoG
Recombinant Endog Fragment (from E. coli)
Catalog #: 10073-124
Supplier:  Prosci
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Size:
    0.05 mg
  • Protein synonyms:
    Endo G|EndoG
  • Protein/peptide name:
    Endog Fragment
  • Purity:
    >95%
  • Molecular weight:
    15 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested applications:
    ELISA, Western Blot
  • Cat. no.:
    10073-124
  • Supplier No.:
    95-103

Specifications

About this item

The fragmentation of nuclear DNA is a hallmark of apoptotic cell death. The activities of caspase and nuclease are involved in the DNA fragmentation. Caspase-activated deoxyribo-nuclease (CAD), also termed DNA fragmentation factor (DFF40), is one such nuclease, and is capable of inducing DNA fragmentation and chromatin condensation after cleavage by caspase-3 of its inhibitor ICAD/DFF45. Caspase and CAD independent DNA fragmentation also exists. Recent studies demonstrated that another nuclease, endonuclease G (EndoG), is specifically activated by apoptotic stimuli and is able to induce nucleosomal fragmentation of DNA independently of caspase and DFF/CAD (1-3). EndoG is a mitochondrion-specific nuclease that translocates to the nucleus and cleaves chromatin DNA during apoptosis. The homologue of mammalian EndoG is the first mitochondrial protein identified to be involved in apoptosis in C. elegans (2). EndoG also cleaves DNA in vitro (4).

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

100mM sodium phosphate, 500mM NaCl, 25 mM imidazole, 10% gycerol.

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought