Order Entry
ContactUsLinkComponent
Mouse Recombinant GDF5 (from E. coli)
Mouse Recombinant GDF5 (from E. coli)
Catalog #: 10073-062
Supplier:  Prosci
CAS Number:  
Mouse Recombinant GDF5 (from E. coli)
Catalog #: 10073-062
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.01 mg
  • Biological activity:
    Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein synonyms:
    OS5|GDF-5|SYNS2|Cartilage-derived morphogenetic protein 1|BDA1C|LAP4|CDMP1|SYM1B|BMP14
  • Protein/peptide name:
    GDF5
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10073-062
  • Supplier No.:
    40-579

Specifications

About this item

GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Grebe Syndrome, which is characterized by short stature, extra digits, short and deformed extremities, and in Hunter-Thompson type dwarfism. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino-acid signal peptide and a 348 amino-acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease. Recombinant murine GDF-5 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acids.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought