Order Entry
ContactUsLinkComponent
Human Recombinant Chemerin (from E. coli)
Human Recombinant Chemerin (from E. coli)
Catalog #: 10073-018
Supplier:  Prosci
CAS Number:  
Human Recombinant Chemerin (from E. coli)
Catalog #: 10073-018
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to chemoattract human immature dendritic cells using a concentration range of 1.0-100.0 ng/mL. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein synonyms:
    TIG2|Chemerin|Retinoic acid receptor responder protein 2|HP10433
  • Protein/peptide name:
    Chemerin
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10073-018
  • Supplier No.:
    40-551

Specifications

About this item

Chemerin is a secreted chemoattractant protein that can signal through the chemokine like receptor-1 (CMKLR1). It is expressed in various tissues, including white adipose tissue, and circulates in blood as an inactive 143 amino acid precursor protein. Biologically active Chemerin is generated by proteolytic removal of C-terminal residues by several circulating proteases. Chemerin acts as a chemoattractant for cells expressing the CMKLR1 receptor, which includes certain dendritic cells, macrophages, and adipocytes. Recombinant human Chemerin is a 15.6 kDa protein consisting of 135 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought