Order Entry
ContactUsLinkComponent
Human Recombinant Cdnf (from E. coli)
Human Recombinant Cdnf (from E. coli)
Catalog #: 10073-016
Supplier:  Prosci
CAS Number:  
Human Recombinant Cdnf (from E. coli)
Catalog #: 10073-016
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to stimulate the proliferation of rat C6 cells. The expected ED50 for this effect is 15-25 μg/mL. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein synonyms:
    ARMETL1|ARMET-like protein 1|Cerebral dopamine neurotrophic factor
  • Protein/peptide name:
    Cdnf
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10073-016
  • Supplier No.:
    40-550

Specifications

About this item

CDNF is a secreted neurotrophic factor that is expressed in brain, neuronal and certain non-neuronal tissues. It has been shown to promote survival, growth and function of dopamine specific neurons. CDNF and its structural homolog MANF, each contain an N-terminal saposin-like lipid binding domain, and a carboxyl-terminal domain, which is not homologous to previously characterized protein structures. CDNF and MANF can prevent 6-OHDA induced degeneration of dopaminergic neurons by triggering survival pathways in a rat experimental model of Parkinson disease. Recombinant human CDNF is an 18.5 kDa protein consisting of 162 amino acids including 8 cysteine residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought