Order Entry
ContactUsLinkComponent
Human Recombinant Activin A (from E. coli)
Human Recombinant Activin A (from E. coli)
Catalog #: 10073-008
Supplier:  Prosci
CAS Number:  
Human Recombinant Activin A (from E. coli)
Catalog #: 10073-008
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological activity:
    Biological activity was determined by its ability to inhibit the proliferation of mouse MPC-11 cells. The expected ED50 is ≤ 2.0 ng/mL, corresponding to a specific activity of ≥ 5 x 105 units/mg. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein synonyms:
    Mothers against decapentaplegic homolog 2|hMAD-2|MAD homolog 2|hSMAD2|JV18-1|MADR2|MADH2|JV18
  • Protein/peptide name:
    Activin A
  • Purity:
    >97%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10073-008
  • Supplier No.:
    40-544

Specifications

About this item

Activin A is a TGF-β family member that exhibits a wide range of biological activities including regulation of cellular proliferation and differentiation, and promotion of neuronal survival. Elevated levels of Activin A in human colorectal tumors and in postmenopausal women have been implicated in colorectal and breast cancers, respectively. The biological activities of Activin A can be neutralized by inhibins and by the diffusible TGF-β antagonist, follistatin. Recombinant human Activin A is a 26.0 kDa disulfide-linked homodimer of two βA chains, each containing 116 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought