To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.002 mg
- Protein synonyms:GMF-beta|GMF|Glia maturation factor beta
- Protein/peptide name:GMFB
- Purity:>98%
- Formulation:Lyophilized
- Cat. no.:10072-602
- Supplier No.:40-508
Specifications
About this item
GMF-β a brain-specific protein that belongs to the actin-binding proteins (ADF) structural family. GMF-β appears to play a role in the differentiation, maintenance, and regeneration of the nervous system. It also supports the progression of certain auto-immune diseases, possibly through its ability to induce the production and secretion of various pro-inflammatory cytokines. Recombinant human GMF-β is a 16.5 kDa globular protein containing 141 amino acid residues including one intramolecular disulfide bond.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.