To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.005 mg
- Biological activity:Biological activity was determined by a cell proliferation using rat C6 cells. The ED50 for this effect is 20.0 - 30.0 ng/mL.
- Protein synonyms:NRN|dJ380B8.2|Neuritin
- Protein/peptide name:Neuritin
- Purity:>98%
- Formulation:Lyophilized
- Cat. no.:10072-600
- Supplier No.:40-507
Specifications
About this item
Neuritin is a neurotrophic factor, which is expressed in response to induction of neuronal activity by NGF, BDNF, NT3, and other neural stimulators. It is expressed primarily in postmitotic-differentiating neurons of the developing nervous system and in neuronal structures related to synaptic plasticity in the adult nervous system. Neuritin acts as a molecular mediator of neurite outgrowth, neuronal survival, and synaptic maturation. Recombinant human Neuritin is a covalently disulfide-linked homodimer, consisting of two 9.7 kDa polypeptide monomers, each containing 88 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.