Order Entry
ContactUsLinkComponent
Human Recombinant IL19 (from E. coli)
Human Recombinant IL19 (from E. coli)
Catalog #: 10072-844
Supplier:  Prosci
CAS Number:  
Human Recombinant IL19 (from E. coli)
Catalog #: 10072-844
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.002 mg
  • Biological activity:
    Biological activity was determined by its ability to activate STAT following receptor - ligand interaction.
  • Protein synonyms:
    NG.1|IL-10C|Melanoma differentiation-associated protein-like protein|ZMDA1|IL-19|MDA1|Interleukin-19
  • Protein/peptide name:
    IL19
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-844
  • Supplier No.:
    40-264

Specifications

About this item

IL-19 belongs to the IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences but they are dissimilar in their biological functions. Preliminary data suggests that IL-19 is a proinflammatory cytokine because it up-regulates IL-6 and TNF-α and induces apoptosis through TNF-α. IL-19 signals through the type I IL-20R. Human and murine IL-19 share 71% amino acid sequence identity. Recombinant human IL-19 is a 17.9 kDa protein containing 153 amino acid residues. In solution IL-19 exists predominantly as a non-disulfide-linked dimer.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought