Order Entry
ContactUsLinkComponent
Human Recombinant ICAM1 (from CHO cells)
Human Recombinant ICAM1 (from CHO cells)
Catalog #: 10072-812
Supplier:  Prosci
CAS Number:  
Human Recombinant ICAM1 (from CHO cells)
Catalog #: 10072-812
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological activity:
    ICAM - 1 supported the adhesion of more than 50% PMA - stimulated HSB2 cells to plates coated at 25 ng/mL ICAM - 1 concentration
  • Protein synonyms:
    ICAM-1|BB2|Intercellular adhesion molecule 1|CD54|Major group rhinovirus receptor|P3.58
  • Protein/peptide name:
    ICAM1
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-812
  • Supplier No.:
    40-238

Specifications

About this item

ICAMs are members of the Ig superfamily of calcium-independent transmembrane glycoproteins. ICAM-1 is a ligand for lymphocyte function-associated (LFA) and Mac-1 integrins and the major human rhinovirus receptor. The primary function of ICAM-1 is to provide adhesion between endothelial cells and leukocytes after stress or injury. The human ICAM-1 gene codes for a 505 amino acid transmembrane glycoprotein containing a 29 amino acid cytoplasmic domain, a 23 amino acid transmembrane domain, and a 453 amino acid extracellular domain. Recombinant human ICAM-1 is a 49.5 kDa glycoprotein comprising the extracellular domain (453 amino acid residues) of ICAM-1. Monomeric glycosylated ICAM-1 migrates at an apparent molecular weight of approximately 72.0-80.0 kDa by SDS-PAGE analysis under reducing conditions.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought