Order Entry
ContactUsLinkComponent
Human Recombinant Relaxin 3 (from E. coli)
Human Recombinant Relaxin 3 (from E. coli)
Catalog #: 10072-796
Supplier:  Prosci
CAS Number:  
Human Recombinant Relaxin 3 (from E. coli)
Catalog #: 10072-796
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Protein synonyms:
    ZINS4|Insulin-like Peptide 7|UNQ6188/PRO20213|Insulin-like Peptide INSL7|H3|INSL7|RXN3|Relaxin-3|insl7
  • Protein/peptide name:
    Relaxin 3
  • Purity:
    >98%
  • Formulation:
    lyophilized
  • Cat. no.:
    10072-796
  • Supplier No.:
    40-230

Specifications

About this item

Relaxin-3 (H3 relaxin, Insulin-like peptide-7, INSL7) is a secreted protein structurally related to insulin, which is expressed primarily in the brain and central nervous system. Relaxin-3 has been identified as the ligand for the GPCR135 receptor, previously known as "somatostatin-like” or "angiotensin-like” peptide receptor, and also binds specifically to the LGR7 receptor, previously identified as an "orphan” G protein coupled receptor. Signaling by Relaxin-3 through its target receptors is, most likely, part of a CNS processing system, activated in response to signaling by neuropeptides and other factors. Intracerebroventricular injections of Relaxin-3 have been shown to cause a significant increase of food intake and body weight in Wistar rats. Recombinant Relaxin-3 is a 5.5 kDa disulfide linked heterodimeric protein consisting of a 24 amino acid A-chain and a 27 amino acid B-chain.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought