Order Entry
ContactUsLinkComponent
Human Recombinant MIA (from E. coli)
Human Recombinant MIA (from E. coli)
Catalog #: 10072-788
Supplier:  Prosci
CAS Number:  
Human Recombinant MIA (from E. coli)
Catalog #: 10072-788
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Gene ID:
    8190
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    The ED50 was determined by the dose - dependant proliferation of the human A375 melanoma cell line. The expected ED50 for this effect is 4 - 6 μg/mL.
  • Protein synonyms:
    CD-RAP|Melanoma inhibitory activity protein|Melanoma-derived growth regulatory protein
  • Protein/peptide name:
    MIA
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-788
  • Supplier No.:
    40-226

Specifications

About this item

MIA is the first discovered member of a family of secreted cytokines termed the MIA/OTOR family. The four known members of this family; MIA, MIA2, OTOR and TANGO each contain a Src homology-3 (SH3)-like domain. MIA is an autocrine growth regulatory protein secreted from chondrocytes and malignant melanoma cells that promotes melanoma metastasis by binding competitively to fibronectin and laminin in a manner that results in melanoma cell detachment from the extracellular matrix in vivo. Elevated levels of MIA may represent a clinically useful marker for diagnosis of melanoma metastasis as well as a potential marker for rheumatoid arthritis. Recombinant human MIA is a 12.2 kDa globular protein containing 108 amino acid residues including two intramolecular disulfide bonds.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought