To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.005 mg
- Biological activity:Biological activity was determined by it's ability to chemoattract activated lymphocytes using a concentration of 1.0 - 100.0 ng/mL.
- Protein synonyms:SR-PSOX|SCYB16|SRPSOX|UNQ2759/PRO6714|CXCLG16|C-X-C motif chemokine 16|Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein
- Protein/peptide name:CXCL16
- Purity:>98%
- Formulation:Lyophilized
- Cat. no.:10072-378
- Supplier No.:40-403
Specifications
About this item
CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal mucin-like stalk sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog). Recombinant human CXCL16 is a 10.1 kDa protein containing 89 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.