To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.005 mg
- Biological activity:Biological activity was determined by its ability to chemoattract human CCR3/HEK 293 cells. The maximum activity was achieved at 2μg/mL.
- Protein synonyms:C-C motif chemokine 26|TSC-1|MIP-4alpha|MIP-4a|UNQ216/PRO242|MIP-4-alpha|IMAC|CC chemokine IMAC|SCYA26
- Protein/peptide name:Eotaxin 3
- Purity:>98%
- Formulation:Lyophilized
- Cat. no.:10073-002
- Supplier No.:40-400
Specifications
About this item
Eotaxin-3 is a CC chemokine that signals through the CCR3 receptor. It is produced by endothelial cells stimulated with IL-4 or IL-13. Eotaxin-3 selectively targets cells expressing CCR3, including eosinophils, basophils, T cells and monocytes. Eotaxin-3 has similar activity to Eotaxin and Eotaxin-2, but the three Eotaxins share only a low degree of sequence homology. Recombinant human Eotaxin-3 is an 8.4 kDa protein containing 71 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.