Order Entry
ContactUsLinkComponent
Human Recombinant SHH (from E. coli)
Human Recombinant SHH (from E. coli)
Catalog #: 10072-732
Supplier:  Prosci
CAS Number:  
Human Recombinant SHH (from E. coli)
Catalog #: 10072-732
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to induce alkaline phosphatase production by C3H/10T1/2 (CCL - 226) cells. The expected ED50 for this effect is 0.8 - 1.0 μg/mL.
  • Protein synonyms:
    TPT|SHH|HLP3|HHG1|SMMCI|HPE3|Sonic hedgehog protein|MCOPCB5|TPTPS|HHG-1
  • Protein/peptide name:
    SHH
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-732
  • Supplier No.:
    40-188

Specifications

About this item

Members of the Hedgehog (Hh) family are highly conserved proteins which are widely represented throughout the animal kingdom. The three known mammalian Hh proteins, Sonic (Shh), Desert (Dhh) and Indian (Ihh) are structurally related and share a high degree of amino-acid sequence identity (e.g., Shh and Ihh are 93% identical). The biologically active form of Hh molecules is obtained by autocatalytic cleavage of their precursor proteins and corresponds to approximately the N-terminal one half of the precursor molecule. Although Hh proteins have unique expression patterns and distinct biological roles within their respective regions of secretion, they use the same signaling pathway and can substitute for each other in experimental systems. Recombinant E. coli derived Human Sonic HedgeHog is a 20.0 kDa protein consisting of 176 amino acid residues, including an N-terminal Ile-Val-Ile sequence substituted for the natural occurring chemically modified Cys residue.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought