To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.005 mg
- Biological activity:The ED50 was determined by the dose - dependent stimulation of thymidine uptake by BaF3 cells expressing FGF - receptors is ≤ 0.5 ng/mL, corresponding to a specific activity of ≥ 2 x 106 units/mg.
- Protein synonyms:FGF-16|MF4|Fibroblast growth factor 16
- Protein/peptide name:FGF16
- Purity:>95%
- Formulation:Lyophilized
- Cat. no.:10072-714
- Supplier No.:40-175
Specifications
About this item
FGF-16 is a heparin binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system. Recombinant human FGF-16 is a 23.7 kDa protein consisting of 207 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.