Human CRTC2 Peptide
Catalog #: 10062-470
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:CRTC2 Peptide
- Concentration:200 ug/mL
- Formulation:Liquid
- Cat. no.:10062-470
- Supplier No.:7325P
Specifications
About this item
16 amino acid peptide near the carboxy terminus of human CRTC2.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
Customers Who Bought This Also Bought