Human SLC27A6 Peptide
Catalog #: 10062-464
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:SLC27A6 Peptide
- Concentration:200 ug/mL
- Formulation:Liquid
- Cat. no.:10062-464
- Supplier No.:7297P
Specifications
About this item
16 amino acid peptide near the amino terminus of human SLC27A6.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
Customers Who Bought This Also Bought