Order Entry
ContactUsLinkComponent
Human Recombinant SCGF-alpha
Human Recombinant SCGF-alpha
Catalog #: 10072-702
Supplier:  Prosci
CAS Number:  
Human Recombinant SCGF-alpha
Catalog #: 10072-702
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    0.002 mg
  • Biological activity:
    The ED50 was determined by TGF - beta1's ability to inhibit the mouse IL - 4 - dependent proliferation of mouse HT - 2 cells is ≤ 0.05 ng/mL, corresponding to a specific activity of ≥ 2 x 107 units/mg.
  • Protein synonyms:
    C-type lectin domain family 11 member A|C-type lectin superfamily member 3|P47|LSLCL|CLECSF3|SCGF
  • Protein/peptide name:
    SCGF-alpha
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-702
  • Supplier No.:
    40-168

Specifications

About this item

The three mammalian isoforms of TGF-β, TGF-β1, β2, β3, signal through the same receptor and elicit similar biological responses. They are multifunctional cytokines that regulate cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix. They are involved in various physiological processes including embryogenesis, tissue remodeling and wound healing. They are secreted predominantly as latent complexes which are stored at the cell surface and in the extracellular matrix. The release of biologically active TGF-β isoform from a latent complex involves proteolytic processing of the complex and /or induction of conformational changes by proteins such as thrombospondin-1. TGF-β1 is the most abundant isoform secreted by almost every cell type. It was originally identified for its ability to induce phenotypic transformation of fibroblasts and recently it has been implicated in the formation of skin tumors. Human TGF-β1 is a 25.0 kDa protein with each subunit containing 112 amino acid residues, linked by a single disulfide bond.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought