Order Entry
ContactUsLinkComponent
Human Recombinant VEGFD (from HEK293 Cells)
Human Recombinant VEGFD (from HEK293 Cells)
Catalog #: 10072-700
Supplier:  Prosci
CAS Number:  
Human Recombinant VEGFD (from HEK293 Cells)
Catalog #: 10072-700
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.002 mg
  • Protein synonyms:
    c-Fos-induced growth factor|VEGF-D|VEGFD|Vascular endothelial growth factor D
  • Protein/peptide name:
    VEGFD
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-700
  • Supplier No.:
    40-166

Specifications

About this item

VEGF-D, a member of the VEGF/PDGF family of structurally related proteins, is a potent angiogenic cytokine. It promotes endothelial cell growth, promotes lymphangiogesis, and can also affect vascular permeability. VEGF-D is highly expressed in the lung, heart, small intestine and fetal lung, and at lower levels in the skeletal muscle, colon, and pancreas. It forms cell surfaced-associated non-covalent disulfide linked homodimers, and can bind and activate both VEGFR-2 (flk1) and VEGFR-3 (flt4) receptors. During embryogenesis, VEGF-D may play a role in the formation of the venous and lymphatic vascular systems. It also participates in the growth and maintenance of differentiated lymphatic endothelium in adults. Both VEGF-C and VEGF-D are over-expressed in certain cancers, and the resulting elevated levels of VEGF-C or VEGF-D tend to correlate with increased lymphatic metastasis. Recombinant human VEGF-D is a 13.1 kDa non-disulfide linked homodimeric protein consisting of two 117 amino acid polypeptide chains. Due to glycosylation the protein migrates as a 20.0-22.0 kDa band under non-reducing condition.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought