Human Nucleobindin-2 Peptide, ProSci
Catalog #: 10062-428
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:Nucleobindin-2 Peptide
- Concentration:200 ug/mL
- Formulation:Liquid
- Cat. no.:10062-428
- Supplier No.:7271P
Specifications
About this item
16 amino acids near the center of human Nucleobindin-2.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
Customers Who Bought This Also Bought