Order Entry
ContactUsLinkComponent
Mouse Recombinant PF4 (from E. coli)
Mouse Recombinant PF4 (from E. coli)
Catalog #: 10072-970
Supplier:  Prosci
CAS Number:  
Mouse Recombinant PF4 (from E. coli)
Catalog #: 10072-970
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to chemoattract human neutrophils using a concentration range of 10.0 - 100.0 ng/mL.
  • Protein synonyms:
    Cxcl4|C-X-C motif chemokine 4|PF-4|Platelet factor 4|Scyb4
  • Protein/peptide name:
    PF4
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-970
  • Supplier No.:
    40-361

Specifications

About this item

PF-4 is a CXC chemokine that is expressed in megakaryocytes and stored in the α-granules of platelets. PF-4 is chemotactic towards neutrophils and monocytes and has been shown to inhibit angiogenesis. Recombinant murine PF-4 is an 8.2 kDa protein containing 76 amino acid residues, including the four highly conserved residues present in CXC chemokines.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought