Order Entry
ContactUsLinkComponent
Mouse Recombinant MIP1B (from E. coli)
Mouse Recombinant MIP1B (from E. coli)
Catalog #: 10072-130
Supplier:  Prosci
CAS Number:  
Mouse Recombinant MIP1B (from E. coli)
Catalog #: 10072-130
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.002 mg
  • Biological activity:
    Biological activity was determined by its ability to chemoattract human monocytes using a concentration range of 20.0 - 100.0 ng/mL.
  • Protein synonyms:
    MIP1 beta
  • Protein/peptide name:
    MIP1B
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-130
  • Supplier No.:
    40-358

Specifications

About this item

Both MIP-1 alpha and MIP-1 beta are structurally and functionally related CC chemokines. They participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells e.g. macrophages, lymphocytes and NK cells. While both MIP-1 alpha and MIP-1 beta exert similar effects on monocytes their effect on lymphocytes differ; with MIP-1 alpha selectively attracting CD8+ lymphocytes and MIP-1 beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1 alpha and MIP-1 beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1 alpha and MIP-1 beta are active on human and murine hematopoietic cells. Recombinant murine MIP-1 beta is a 7.8 kDa protein containing 69 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought