To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:0.005 mg
- Biological activity:Biological activity was determined by its ability to chemoattract murine lymphocytes using a concentration range of 1.0 - 10.0 ng/mL.
- Protein synonyms:MEC|Small-inducible cytokine A28|C-C motif chemokine 28|CCK1|Scya28
- Protein/peptide name:MEC
- Purity:>98%
- Formulation:Lyophilized
- Cat. no.:10072-966
- Supplier No.:40-357
Specifications
About this item
MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines including the four highly conserved cysteine residues present in CC chemokines. Recombinant murine MEC is a 12.6 kDa protein containing 111 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.