Order Entry
ContactUsLinkComponent
Human Recombinant Epiregulin (from E. coli)
Human Recombinant Epiregulin (from E. coli)
Catalog #: 10072-682
Supplier:  Prosci
CAS Number:  
Human Recombinant Epiregulin (from E. coli)
Catalog #: 10072-682
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    The ED50 was determined by its ability to inhibit IGF - II induced proliferation of MCF - 7 is ≤ 0.3 μg/mL in the presence of 15 ng/mL of human IGF - II.
  • Protein synonyms:
    Proepiregulin|ER|EPR
  • Protein/peptide name:
    Epiregulin
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-682
  • Supplier No.:
    40-149

Specifications

About this item

Epiregulin is an EGF related growth factor that binds specifically to EGFR (ErbB1) and ErbB4, but not ErbB2 or ErbB3. It is expressed mainly in the placenta and peripheral blood leukocytes and in certain carcinomas of the bladder, lung, kidney and colon. Epiregulin stimulates the proliferation of keratinocytes, hepatocytes, fibroblasts and vascular smooth muscle cells. It also inhibits the growth of several tumor-derived epithelial cell lines. Human Epiregulin is initially synthesized as a glycosylated 19.0 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a 6.0 kDa mature secreted sequence. Recombinant human Epiregulin is a 5.6 kDa monomeric protein, containing 50 amino residues, which corresponds to the mature secreted Epiregulin sequence.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought